missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-hydroxypyrazole, 98%

Catalog No. AC271720050
Change view
Click to view available options
Quantity:
5 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC271720050 5 g Glass bottle
AC271721000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC271720050 Supplier Thermo Scientific Chemicals Supplier No. 271720050
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6126-22-3 | C3H5N3O | 99.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6126-22-3
MDL Number MFCD00022384
InChI Key QZBGOTVBHYKUDS-UHFFFAOYSA-N
Synonym 3-amino-5-hydroxypyrazole, 3-amino-1h-pyrazol-5-ol, 5-amino-1h-pyrazol-3-ol, 5-amino-1h-pyrazol-3 2h-one, 3-amino-1,2-dihydropyrazol-5-one, 5-aminopyrazol-3-ol, 5-amino-2h-pyrazol-3-ol, 3-pyrazolin-5-one, 3-amino, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 1h-pyrazol-5-ol, 3-amino
PubChem CID 96221
IUPAC Name 5-amino-1,2-dihydropyrazol-3-one
SMILES C1=C(NNC1=O)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Beige-Brown to Beige-Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C3H5N3O
Quantity 5 g
Beilstein 25, 3516
Molecular Weight (g/mol) 99.09
Formula Weight 99.09
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-hydroxypyrazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 228-095-6

RUO – Research Use Only