missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-hydroxy-1H-pyrazole, 98%

CAS: 6126-22-3 | C3H5N3O | 99.093 g/mol

Supplier:  Thermo Scientific Chemicals A1141214

 View more versions of this product

Catalog No. AAA1141214

Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

3-Amino-5-hydroxy-1H-pyrazole
∼210°C (decomposition)
C3H5N3O
25 g
3-amino-5-hydroxypyrazole, 3-amino-1h-pyrazol-5-ol, 5-amino-1h-pyrazol-3-ol, 5-amino-1h-pyrazol-3 2h-one, 3-amino-1,2-dihydropyrazol-5-one, 5-aminopyrazol-3-ol, 5-amino-2h-pyrazol-3-ol, 3-pyrazolin-5-one, 3-amino, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 1h-pyrazol-5-ol, 3-amino
C1=C(NNC1=O)N
99.093
99.09
6126-22-3
Odorless
MFCD00022384
507793
QZBGOTVBHYKUDS-UHFFFAOYSA-N
5-amino-1,2-dihydropyrazol-3-one
96221
98%

Chemical Identifiers

6126-22-3
99.093
QZBGOTVBHYKUDS-UHFFFAOYSA-N
96221
C1=C(NNC1=O)N
C3H5N3O
MFCD00022384
3-amino-5-hydroxypyrazole, 3-amino-1h-pyrazol-5-ol, 5-amino-1h-pyrazol-3-ol, 5-amino-1h-pyrazol-3 2h-one, 3-amino-1,2-dihydropyrazol-5-one, 5-aminopyrazol-3-ol, 5-amino-2h-pyrazol-3-ol, 3-pyrazolin-5-one, 3-amino, 5-amino-2,3-dihydro-1h-pyrazol-3-one, 1h-pyrazol-5-ol, 3-amino
5-amino-1,2-dihydropyrazol-3-one
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

EINECSNumber : 228-095-6

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only