missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-fluoropyridine 97.0+%, TCI America™
$104.38 - $354.17
Chemical Identifiers
| CAS | 210169-05-4 |
|---|---|
| Molecular Formula | C5H5FN2 |
| Molecular Weight (g/mol) | 112.107 |
| MDL Number | MFCD00234073 |
| InChI Key | ZRORIJXOWXYPMO-UHFFFAOYSA-N |
| Synonym | 3-amino-5-fluoropyridine, 5-amino-3-fluoropyridine, 3-pyridinamine, 5-fluoro, 5-fluoro-3-pyridylamine, 5-fluoro-3-pyridinamine, 3-fluoro-5-aminopyridine, 5-fluoropyridin-3-ylamine, 3-amine-5-fluoro-pyridin, 3-amino-5-fluoro-pyridin, 3-amine-5-fluoro-pyridine |
| PubChem CID | 574450 |
| IUPAC Name | 5-fluoropyridin-3-amine |
| SMILES | C1=C(C=NC=C1F)N |
Chemical Identifiers
| 210169-05-4 | |
| 112.107 | |
| ZRORIJXOWXYPMO-UHFFFAOYSA-N | |
| 574450 | |
| C1=C(C=NC=C1F)N |
| C5H5FN2 | |
| MFCD00234073 | |
| 3-amino-5-fluoropyridine, 5-amino-3-fluoropyridine, 3-pyridinamine, 5-fluoro, 5-fluoro-3-pyridylamine, 5-fluoro-3-pyridinamine, 3-fluoro-5-aminopyridine, 5-fluoropyridin-3-ylamine, 3-amine-5-fluoro-pyridin, 3-amino-5-fluoro-pyridin, 3-amine-5-fluoro-pyridine | |
| 5-fluoropyridin-3-amine |
Specifications
| 210169-05-4 | |
| Yellow | |
| MFCD00234073 | |
| 3-amino-5-fluoropyridine, 5-amino-3-fluoropyridine, 3-pyridinamine, 5-fluoro, 5-fluoro-3-pyridylamine, 5-fluoro-3-pyridinamine, 3-fluoro-5-aminopyridine, 5-fluoropyridin-3-ylamine, 3-amine-5-fluoro-pyridin, 3-amino-5-fluoro-pyridin, 3-amine-5-fluoro-pyridine | |
| C1=C(C=NC=C1F)N | |
| 112.107 | |
| 112.11 | |
| Crystalline Powder |
| 87°C | |
| C5H5FN2 | |
| 1 g | |
| ZRORIJXOWXYPMO-UHFFFAOYSA-N | |
| 5-fluoropyridin-3-amine | |
| 574450 | |
| ≥97.0% (GC,T) | |
| 3-Amino-5-fluoropyridine |
Safety and Handling
TSCA : No