missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-5-bromobenzotrifluoride 98.0+%, TCI America™
$82.02 - $246.05
Chemical Identifiers
| CAS | 54962-75-3 |
|---|---|
| Molecular Formula | C7H5BrF3N |
| Molecular Weight (g/mol) | 240.023 |
| InChI Key | HJTLKVYOWNTDPF-UHFFFAOYSA-N |
| Synonym | 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine |
| PubChem CID | 2735880 |
| IUPAC Name | 3-bromo-5-(trifluoromethyl)aniline |
| SMILES | C1=C(C=C(C=C1N)Br)C(F)(F)F |
Chemical Identifiers
| 54962-75-3 | |
| 240.023 | |
| 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine | |
| 3-bromo-5-(trifluoromethyl)aniline |
| C7H5BrF3N | |
| HJTLKVYOWNTDPF-UHFFFAOYSA-N | |
| 2735880 | |
| C1=C(C=C(C=C1N)Br)C(F)(F)F |
Specifications
| 54962-75-3 | |
| C7H5BrF3N | |
| 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine | |
| C1=C(C=C(C=C1N)Br)C(F)(F)F | |
| 240.023 | |
| 240.02 | |
| Liquid |
| 223°C | |
| 5 g | |
| HJTLKVYOWNTDPF-UHFFFAOYSA-N | |
| 3-bromo-5-(trifluoromethyl)aniline | |
| 2735880 | |
| ≥98.0% (GC) | |
| 3-Amino-5-bromobenzotrifluoride |
Safety and Handling
TSCA : No