missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromobenzotrifluoride, 97%

Catalog No. AC442660050
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC442660050 5 g
AC442660250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC442660050 Supplier Thermo Scientific Chemicals Supplier No. 442660050

CAS: 54962-75-3 | C7H5BrF3N | 240.02 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 54962-75-3
Molecular Formula C7H5BrF3N
Molecular Weight (g/mol) 240.02
InChI Key HJTLKVYOWNTDPF-UHFFFAOYSA-N
Synonym 3-amino-5-bromobenzotrifluoride, 3-bromo-5-trifluoromethyl aniline, 3-bromo-5-trifluoromethyl-phenylamine, 3-amino-5-bromo trifluoromethyl benzene, 3-bromo-5-trifluoromethyl benzenamine, 3-bromo-5-aminobenzotrifluoride, 3-amino-5-bromotrifluorotoluene, benzenamine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluoromethyl phenylamine
PubChem CID 2735880
IUPAC Name 3-bromo-5-(trifluoromethyl)aniline
SMILES C1=C(C=C(C=C1N)Br)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.6970g/mL
Boiling Point 220.0°C to 223.0°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Quantity 5 g
Specific Gravity 1.697
Formula Weight 240.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Amino-5-bromobenzotrifluoride
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 259-412-6

RUO – Research Use Only