Learn More
3-Amino-5-bromo-1H-indazole, 95%, Thermo Scientific™
Chemical Identifiers
| CAS | 61272-71-7 |
|---|---|
| Molecular Formula | C7H6BrN3 |
| Molecular Weight (g/mol) | 212.05 |
| MDL Number | MFCD03426696 |
| InChI Key | OMPYFDJVSAMSMA-UHFFFAOYSA-N |
| Synonym | 3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine |
| PubChem CID | 817910 |
| IUPAC Name | 5-bromo-1H-indazol-3-amine |
| SMILES | NC1=NNC2=CC=C(Br)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC435760010
|
N/A | 1g |
N/A
|
|
|||||
|
AC435760050
|
N/A | 5g |
N/A
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 61272-71-7 | |
| 212.05 | |
| OMPYFDJVSAMSMA-UHFFFAOYSA-N | |
| 817910 | |
| NC1=NNC2=CC=C(Br)C=C12 |
| C7H6BrN3 | |
| MFCD03426696 | |
| 3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine | |
| 5-bromo-1H-indazol-3-amine |
Specifications
| 61272-71-7 | |
| 100.0 | |
| 94% min. (HPLC) | |
| C7H6BrN3 | |
| 3-amino-5-bromo-1h-indazole, 3-amino-5-bromoindazole, 5-bromo-1h-indazol-3-ylamine, 1h-indazol-3-amine, 5-bromo, 5-bromo-1h-indazole-3-ylamine, pubchem18573, 3-amino-5-bromo-indazole, acmc-1b3h9, 5-bromo-1h-indazole-3-amine | |
| NC1=NNC2=CC=C(Br)C=C12 | |
| 212.05 | |
| 212.05 | |
| 3-Amino-5-bromo-1H-indazole |
| 94.0 | |
| Authentic | |
| Glass bottle | |
| MFCD03426696 | |
| OMPYFDJVSAMSMA-UHFFFAOYSA-N | |
| 5-bromo-1H-indazol-3-amine | |
| 817910 | |
| 95% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for
GHS Signal Word: Warning
RUO – Research Use Only