accessibility menu, dialog, popup

UserName

BrNHNH2N

CAS RN 61272-71-7

IUPAC Name: 5-bromo-1H-indazol-3-amine
Synonyms: 5-bromo-1H-indazol-3-amine
Molecular Weight (g/mol): 212.05
Molecular Formula: C7H6BrN3
InChi Key: OMPYFDJVSAMSMA-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=C(Br)C=C12
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results