missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-5-bromo-1-methyl-1H-indazole, 97%

Catalog No. p-7031928
Change view
Click to view available options
Quantity:
250 mg
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH33202MD 250 mg
AAH3320206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH33202MD Supplier Thermo Scientific Chemicals Supplier No. H33202MD
Only null left

CAS: 1000018-06-3 | C8H8BrN3 | 226.077 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1000018-06-3
Molecular Formula C8H8BrN3
Molecular Weight (g/mol) 226.077
MDL Number MFCD09864804
InChI Key BKBSBRJIGMVBFM-UHFFFAOYSA-N
Synonym 5-bromo-1-methyl-1h-indazol-3-amine, 3-amino-5-bromo-1-methyl-1h-indazole, acmc-2097ln, 5-bromo-1-methyl-1h-indazole-3-ylamine
PubChem CID 26369215
IUPAC Name 5-bromo-1-methylindazol-3-amine
SMILES CN1C2=C(C=C(C=C2)Br)C(=N1)N

Specifications

Melting Point 133°C to 135°C
Quantity 250 mg
Formula Weight 226.08
Percent Purity 97%
Chemical Name or Material 3-Amino-5-bromo-1-methyl-1H-indazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only