CAS RN 1000018-06-3
IUPAC Name:
5-bromo-1-methyl-1H-indazol-3-amine
Synonyms:
5-bromo-1-methylindazol-3-amine
Molecular Weight (g/mol):
226.08
Molecular Formula:
C8H8BrN3
InChi Key:
BKBSBRJIGMVBFM-UHFFFAOYSA-N
SMILES:
CN1N=C(N)C2=CC(Br)=CC=C12
1
–
2
of
2
results
1
Promotions Available
3-Amino-5-bromo-1-methyl-1H-indazole, 97%, Thermo Scientific™
3-Amino-5-bromo-1-methyl-1H-indazole, 97%, Thermo Scientific™
2
Promotions Available
3-Amino-5-bromo-1-methyl-1H-indazole, 97%
3-Amino-5-bromo-1-methyl-1H-indazole, 97%
CAS: 1000018-06-3 Molecular Formula: C8H8BrN3 Molecular Weight (g/mol): 226.077 MDL Number: MFCD09864804 InChI Key: BKBSBRJIGMVBFM-UHFFFAOYSA-N Synonym: 5-bromo-1-methyl-1h-indazol-3-amine,3-amino-5-bromo-1-methyl-1h-indazole,acmc-2097ln,5-bromo-1-methyl-1h-indazole-3-ylamine PubChem CID: 26369215 IUPAC Name: 5-bromo-1-methylindazol-3-amine SMILES: CN1C2=C(C=C(C=C2)Br)C(=N1)N