missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-phenyl-1H-pyrazole, 95%

Catalog No. AAH2589503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH2589503 1 g
AAH2589506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH2589503 Supplier Thermo Scientific Chemicals Supplier No. H2589503

CAS: 5591-70-8 | C9H9N3 | 159.192 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5591-70-8
Molecular Formula C9H9N3
Molecular Weight (g/mol) 159.192
MDL Number MFCD01693715
InChI Key QEHKQNYBBLCFIJ-UHFFFAOYSA-N
Synonym 4-phenyl-1h-pyrazol-3-amine, 3-amino-4-phenylpyrazole, 3-amino-4-phenyl-1h-pyrazole, 4-phenyl-2h-pyrazol-3-ylamine, usaf el-34, 3-amino-4-phenyl pyrazole, 4-phenyl-1h-pyrazol-3-ylamine, 1h-pyrazol-4-amine, 4-phenyl-9ci, 4-phenyl-2h-pyrazol-3-amine, pyrazole, 3-amino-4-phenyl
PubChem CID 79703
IUPAC Name 4-phenyl-1H-pyrazol-5-amine
SMILES C1=CC=C(C=C1)C2=C(NN=C2)N

Specifications

Melting Point 170°C to 173°C
Quantity 1 g
Formula Weight 159.19
Percent Purity 95%
Chemical Name or Material 3-Amino-4-phenyl-1H-pyrazole
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only