missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-4-methylbenzoic Acid 98.0+%, TCI America™
$115.57 - $866.79
Chemical Identifiers
| CAS | 2458-12-0 |
|---|---|
| Molecular Formula | C8H9NO2 |
| Molecular Weight (g/mol) | 151.165 |
| MDL Number | MFCD00007742 |
| InChI Key | XKFIFYROMAAUDL-UHFFFAOYSA-N |
| Synonym | 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh |
| PubChem CID | 75568 |
| IUPAC Name | 3-amino-4-methylbenzoic acid |
| SMILES | CC1=C(C=C(C=C1)C(=O)O)N |
Chemical Identifiers
| 2458-12-0 | |
| 151.165 | |
| XKFIFYROMAAUDL-UHFFFAOYSA-N | |
| 75568 | |
| CC1=C(C=C(C=C1)C(=O)O)N |
| C8H9NO2 | |
| MFCD00007742 | |
| 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh | |
| 3-amino-4-methylbenzoic acid |
Specifications
| 2458-12-0 | |
| C8H9NO2 | |
| 25 g | |
| XKFIFYROMAAUDL-UHFFFAOYSA-N | |
| 3-amino-4-methylbenzoic acid | |
| 75568 | |
| ≥98.0% (HPLC,T) | |
| 3-Amino-4-methylbenzoic Acid |
| 166°C | |
| MFCD00007742 | |
| 3-amino-p-toluic acid, 3-amino-4-methyl-benzoic acid, benzoic acid, 3-amino-4-methyl, 3-amino-4-methylbenzoicacid, benzoic acid, 3-amino-4-methyl-9ci, pubchem8194, 3-amino-4-toluic acid, rarechem al bo 1039, h-3 atl 4-oh, h-3 abz 4-me-oh | |
| CC1=C(C=C(C=C1)C(=O)O)N | |
| 151.165 | |
| 151.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No