missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybiphenyl 98.0+%, TCI America™

Catalog No. p-7117156
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A09585G 5 g
A095825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A09585G Supplier TCI America Supplier No. A09585G
Only null left

Chemical Identifiers

CAS 39811-17-1
Molecular Formula C13H13NO
Molecular Weight (g/mol) 199.253
MDL Number MFCD00007790
InChI Key DTYBRSLINXBXMP-UHFFFAOYSA-N
Synonym 5-phenyl-o-anisidine, 3-amino-4-methoxybiphenyl, 4-methoxy-1,1'-biphenyl-3-amine, 2-amino-4-phenylanisole, 4-methoxybiphenyl-3-amine, 1,1'-biphenyl-3-amine, 4-methoxy, 2-methoxy-5-phenylphenylamine, acmc-209j7y, 2-methoxy-5-phenyl-aniline, 3-amino-4-methoxy-biphenyl
PubChem CID 123489
IUPAC Name 2-methoxy-5-phenylaniline
SMILES COC1=C(C=C(C=C1)C2=CC=CC=C2)N

Specifications

Melting Point 84°C
Quantity 5 g
Formula Weight 199.25
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybiphenyl