missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzoic Acid 98.0+%, TCI America™

Catalog No. A19555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A19555G 5 g
A195525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A19555G Supplier TCI America Supplier No. A19555G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 2840-26-8
Molecular Formula C8H9NO3
Molecular Weight (g/mol) 167.164
MDL Number MFCD00002521
InChI Key FDGAEAYZQQCBRN-UHFFFAOYSA-N
Synonym 3-amino-p-anisic acid, p-anisic acid, 3-amino, 3-amino-4-methoxy-benzoic acid, 3-amino-4-anisic acid, 3-amino-4-methoxybenzoicacid, benzoic acid, 3-amino-4-methoxy, kyselina 3-amino-4-methoxybenzoova, 3-amino-4-methoxybenzoic, 5-carboxy-2-methoxyaniline, kyselina 3-amino-4-methoxybenzoova czech
PubChem CID 17823
IUPAC Name 3-amino-4-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)C(=O)O)N

Specifications

Melting Point 205°C
Quantity 5 g
Formula Weight 167.16
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybenzoic Acid
EINECSNumber (3)-1600&(3)-1632
RTECSNumber DG2872000
TSCA No