missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzanilide 98.0+%, TCI America™

Catalog No. A195425G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A195425G 25 g
A1954250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A195425G Supplier TCI America Supplier No. A195425G

Chemical Identifiers

CAS 120-35-4
Molecular Formula C14H14N2O2
Molecular Weight (g/mol) 242.278
MDL Number MFCD00017166
InChI Key LHMQDVIHBXWNII-UHFFFAOYSA-N
Synonym 3-amino-4-methoxybenzanilide, 3-amino-p-anisanilide, benzamide, 3-amino-4-methoxy-n-phenyl, 3-amino-4-methoxy benzanilide, benzanilide, 3-amino-4-methoxy, p-anisanilide, 3-amino, unii-v5y646aw8s, amba, 3-amino-4-methoxy-n-phenyl-benzamide, 3-amino-4-methoxyphenyl-n-benzamide
PubChem CID 8426
IUPAC Name 3-amino-4-methoxy-N-phenylbenzamide
SMILES COC1=C(C=C(C=C1)C(=O)NC2=CC=CC=C2)N

Specifications

Melting Point 156°C
Color White-Yellow
Quantity 25 g
Formula Weight 242.28
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybenzanilide
EINECSNumber (3)-1554
RTECSNumber CV7400000
TSCA Yes