missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-methoxybenzamide 98.0+%, TCI America™

Catalog No. A195325G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A195325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A195325G Supplier TCI America Supplier No. A195325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 17481-27-5
Molecular Formula C8H10N2O2
Molecular Weight (g/mol) 166.18
MDL Number MFCD00017132
InChI Key INCJNDAQNPWMPZ-UHFFFAOYSA-N
Synonym benzamide, 3-amino-4-methoxy, 3-amino-p-anisamide, 3-amino-4-methoxy-benzamide, 3-amino-4-methoxy benzamide, acmc-209e8v, 5-amino-4-methoxybenzamide, cambridge id 5306680, ksc495o3j, incjndaqnpwmpz-uhfffaoysa
PubChem CID 87135
IUPAC Name 3-amino-4-methoxybenzamide
SMILES COC1=C(C=C(C=C1)C(=O)N)N

Specifications

Melting Point 157°C
Quantity 25 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-4-methoxybenzamide
EINECSNumber (3)-1475
TSCA Yes