missing translation for 'onlineSavingsMsg'
Learn More

3'-Amino-4'-methoxyacetanilide 98.0+%, TCI America™

Catalog No. A106425G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A106425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A106425G Supplier TCI America Supplier No. A106425G
Only null left

Chemical Identifiers

CAS 6375-47-9
Molecular Formula C9H12N2O2
Molecular Weight (g/mol) 180.207
MDL Number MFCD00008676
InChI Key SJWQCBCAGCEWCV-UHFFFAOYSA-N
Synonym 5-Acetamido-o-anisidine, N1-Acetyl-4-methoxy-1,3-phenylenediamine
PubChem CID 80779
IUPAC Name N-(3-amino-4-methoxyphenyl)acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)OC)N

Specifications

Melting Point 109°C
Quantity 25 g
Formula Weight 180.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Amino-4′-methoxyacetanilide
EINECSNumber (3)-0731&(3)-2797
TSCA Yes