missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4-hydroxyphenyl Methyl Sulfone 97.0+%, TCI America™

$46.20 - $115.51

Chemical Identifiers

CAS 98-30-6
Molecular Formula C7H9NO3S
Molecular Weight (g/mol) 187.21
MDL Number MFCD00719450
InChI Key SFLMBHYNCSYPOO-UHFFFAOYSA-N
Synonym 2-Amino-4-(methylsulfonyl)phenol, 2-Hydroxy-5-(methylsulfonyl)aniline
PubChem CID 7382
IUPAC Name 2-amino-4-methanesulfonylphenol
SMILES CS(=O)(=O)C1=CC=C(O)C(N)=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
A21985G
View Documents
TCI America
A21985G
5 g
Each for $46.20
Only null left
 
A219825G
View Documents
TCI America
A219825G
25 g
Each for $115.51
Only null left
 
Description

Description

Specifications

Chemical Identifiers

98-30-6
187.21
SFLMBHYNCSYPOO-UHFFFAOYSA-N
7382
CS(=O)(=O)C1=CC=C(O)C(N)=C1
C7H9NO3S
MFCD00719450
2-Amino-4-(methylsulfonyl)phenol, 2-Hydroxy-5-(methylsulfonyl)aniline
2-amino-4-methanesulfonylphenol

Specifications

98-30-6
C7H9NO3S
5 g
SFLMBHYNCSYPOO-UHFFFAOYSA-N
2-amino-4-methanesulfonylphenol
7382
≥97.0% (HPLC,T)
3-Amino-4-hydroxyphenyl Methyl Sulfone
158°C
MFCD00719450
2-Amino-4-(methylsulfonyl)phenol, 2-Hydroxy-5-(methylsulfonyl)aniline
CS(=O)(=O)C1=CC=C(O)C(N)=C1
187.21
187.21
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-2140

TSCA : Yes

SDS
missing translation for 'documents'

missing translation for 'documents'