missing translation for 'onlineSavingsMsg'
Learn More

3'-Amino-4'-fluoroacetophenone 98.0+%, TCI America™

Catalog No. A29501G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A29501G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A29501G Supplier TCI America Supplier No. A29501G
Only null left

Chemical Identifiers

CAS 2002-82-6
Molecular Formula C8H8FNO
Molecular Weight (g/mol) 153.16
MDL Number MFCD00115400
InChI Key LUKSDHOQKVTGGT-UHFFFAOYSA-N
PubChem CID 10606953
IUPAC Name 1-(3-amino-4-fluorophenyl)ethan-1-one
SMILES CC(=O)C1=CC(N)=C(F)C=C1

Specifications

Melting Point 71°C
Color Yellow
Quantity 1 g
Formula Weight 153.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Amino-4′-fluoroacetophenone