accessibility menu, dialog, popup

UserName

CAS RN 2002-82-6

IUPAC Name: 1-(3-amino-4-fluorophenyl)ethan-1-one
Synonyms: 1-(3-amino-4-fluorophenyl)ethanone
Molecular Weight (g/mol): 153.16
Molecular Formula: C8H8FNO
InChi Key: LUKSDHOQKVTGGT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC(N)=C(F)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results