missing translation for 'onlineSavingsMsg'
Learn More

3'-Amino-4'-chloroacetanilide 98.0+%, TCI America™

Catalog No. a155925g
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A155925G 25 g
A15595G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A155925G Supplier TCI America Supplier No. A155925G
Only null left

Chemical Identifiers

CAS 51867-83-5
Molecular Formula C8H9ClN2O
Molecular Weight (g/mol) 184.623
MDL Number MFCD02093411
InChI Key MIIPQGGYCFVDAI-UHFFFAOYSA-N
Synonym n-3-amino-4-chlorophenyl acetamide, 3'-amino-4'-chloroacetanilide, 5-acetylamido-2-chloroaniline, 3-amino-4-chloroacetanilide, n-3-amino-4-chloro-phenyl acetamide, n-3-amino-4-chlorphenyl acetamid, acetamide, n-3-amino-4-chlorophenyl, n-3-azanyl-4-chloranyl-phenyl ethanamide, acmc-209kwg, ksc595a1f
PubChem CID 103996
ChEBI CHEBI:86568
IUPAC Name N-(3-amino-4-chlorophenyl)acetamide
SMILES CC(=O)NC1=CC(=C(C=C1)Cl)N

Specifications

Melting Point 172°C
Quantity 25 g
Physical Form Crystal-Powder at 20°C
Formula Weight 184.62
Percent Purity ≥98.0% (T)
Chemical Name or Material 3′-Amino-4′-chloroacetanilide
EINECSNumber (3)-0275
TSCA No