accessibility menu, dialog, popup

UserName

CAS RN 51867-83-5

CH3ONHClH2N

CAS RN 51867-83-5

IUPAC Name: N-(3-amino-4-chlorophenyl)acetamide
Synonyms: N-(3-amino-4-chlorophenyl)acetamide
Molecular Weight (g/mol): 184.62
Molecular Formula: C8H9ClN2O
InChi Key: MIIPQGGYCFVDAI-UHFFFAOYSA-N
SMILES: CC(=O)NC1=CC=C(Cl)C(N)=C1
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results