missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole, 98+%

Catalog No. AAL0647609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0647609 10 g
AAL0647618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0647609 Supplier Thermo Scientific Chemicals Supplier No. L0647609

CAS: 3314-35-0 | C9H11N3 | 161.208 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3314-35-0
Molecular Formula C9H11N3
Molecular Weight (g/mol) 161.208
MDL Number MFCD00051730
InChI Key QENUTIJJGGTTPE-UHFFFAOYSA-N
Synonym 1-phenyl-4,5-dihydro-1h-pyrazol-3-amine, 2-pyrazoline, 3-amino-1-phenyl, 1-fenyl-3-aminopyrazolin, 3-amino-1-phenyl-2-pyrazoline, 1-phenyl-4,5-dihydro-1h-pyrazol-3-ylamine, 3-amino-4,5-dihydro-1-phenylpyrazole, pyrazolidine, 3-imino-1-phenyl, 4,5-dihydro-1-phenyl-1h-pyrazol-3-amine, 3-amino-1-fenyl-2-pyrazolin, 1-phenyl-3-amino-2-pyrazoline
PubChem CID 98736
IUPAC Name 2-phenyl-3,4-dihydropyrazol-5-amine
SMILES C1CN(N=C1N)C2=CC=CC=C2

Specifications

Melting Point 167°C to 170°C
Quantity 10 g
Beilstein 139162
Formula Weight 161.21
Percent Purity ≥98%
Chemical Name or Material 3-Amino-4,5-dihydro-1-phenyl-1H-pyrazole
Hazard Category H319
Hazard Statement GHS H Statement
H319
Causes serious eye irritation.
Precautionary Statement P264b-P280i-P305+P351+P338
EINECSNumber 222-005-9
RTECSNumber UQ8910000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only