accessibility menu, dialog, popup

UserName

NH2NN

CAS RN 3314-35-0

IUPAC Name: 1-phenyl-4,5-dihydro-1H-pyrazol-3-amine
Synonyms: 1-phenyl-4,5-dihydropyrazol-3-amine
Molecular Weight (g/mol): 161.21
Molecular Formula: C9H11N3
InChi Key: QENUTIJJGGTTPE-UHFFFAOYSA-N
SMILES: NC1=NN(CC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results