missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-phenylpropionic Acid 98.0+%, TCI America™

Supplier:  TCI America A248025G

Quantity 25 g

 View more versions of this product

Catalog No. A248025G


Only null left

Description

Description

Specifications

Chemical Identifiers

614-19-7
165.19
UJOYFRCOTPUKAK-UHFFFAOYNA-N
69189
3-amino-3-phenylpropanoic acid
C9H11NO2
MFCD00008064,MFCD01076238
3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid
CHEBI:68528
NC(CC(O)=O)C1=CC=CC=C1

Specifications

223°C
614-19-7
C9H11NO2
3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid
NC(CC(O)=O)C1=CC=CC=C1
25 g
69189
165.19
Crystalline Powder
3-Amino-3-phenylpropionic Acid
White
MFCD00008064,MFCD01076238
UJOYFRCOTPUKAK-UHFFFAOYNA-N
3-amino-3-phenylpropanoic acid
165.19
CHEBI:68528
≥98.0% (HPLC,T)
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions