missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-phenylpropionic Acid 98.0+%, TCI America™

Catalog No. p-7117933
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24805G 5 g
A248025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24805G Supplier TCI America Supplier No. A24805G

Chemical Identifiers

CAS 614-19-7
Molecular Formula C9H11NO2
Molecular Weight (g/mol) 165.19
MDL Number MFCD00008064,MFCD01076238
InChI Key UJOYFRCOTPUKAK-UHFFFAOYNA-N
Synonym 3-amino-3-phenylpropionic acid, dl-3-amino-3-phenylpropionic acid, dl-beta-phenylalanine, beta-phenyl-beta-alanine, 3-phenyl-dl-beta-alanine, h-dl-beta-phe-oh, benzenepropanoic acid, beta-amino, 3-amino-3-phenyl-propionic acid, 3a3ppa, beta-aminohydrocinnamic acid
PubChem CID 69189
ChEBI CHEBI:68528
IUPAC Name 3-amino-3-phenylpropanoic acid
SMILES NC(CC(O)=O)C1=CC=CC=C1

Specifications

Melting Point 223°C
Color White
Quantity 5 g
Formula Weight 165.19
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-phenylpropionic Acid