missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(4-chlorophenyl)propionic Acid 97.0+%, TCI America™

Catalog No. p-7117901
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A24291G 1 g
A24295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A24291G Supplier TCI America Supplier No. A24291G

Chemical Identifiers

CAS 19947-39-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.634
MDL Number MFCD00126370
InChI Key BXGDBHAMTMMNTO-UHFFFAOYSA-N
Synonym 3-amino-3-4-chlorophenyl propanoic acid, 3-amino-3-4-chlorophenyl propionic acid, 3-p-chlorophenyl-dl-beta-alanine, 3-amino-3-4-chloro-phenyl-propionic acid, dl-beta-p-chlorophenyl alanine, 3-amino-3-p-chlorophenyl propionic acid, dl-3-amino-3-4-chlorophenyl-propionic acid, dl-beta-4-chlorophenyl alanine, rac-3-amino-3-4-chlorophenyl propionic acid, dl-3-amino-3-4'-chlorophenyl propionic acid
PubChem CID 344176
IUPAC Name 3-amino-3-(4-chlorophenyl)propanoic acid
SMILES C1=CC(=CC=C1C(CC(=O)O)N)Cl

Specifications

Melting Point 220°C
Color White
Quantity 1 g
Formula Weight 199.63
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-3-(4-chlorophenyl)propionic Acid