missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-pyridone 96.0+%, TCI America™

Catalog No. A22855G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22855G 5 g
A22851G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22855G Supplier TCI America Supplier No. A22855G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 33630-99-8 | C5H6N2O | 110.12 g/mol

Chemical Identifiers

CAS 33630-99-8
Molecular Formula C5H6N2O
Molecular Weight (g/mol) 110.12
MDL Number MFCD03411556
InChI Key VTSFNCCQCOEPKF-UHFFFAOYSA-N
Synonym 3-amino-2-hydroxypyridine, 3-aminopyridin-2-ol, 3-amino-2-pyridinol, 3-aminopyridin-2 1h-one, 2 1h-pyridinone, 3-amino, 3-amino-2-pyridone, 3-amino-pyridin-2-ol, 2-pyridinol, 3-amino, 2-hydroxy-3-aminopyridine, 3-aminohydropyridin-2-one
PubChem CID 322353
IUPAC Name 3-amino-1,2-dihydropyridin-2-one
SMILES NC1=CC=CNC1=O

Specifications

Melting Point 124°C
Quantity 5 g
Formula Weight 110.12
Percent Purity ≥96.0% (GC,T)
Grade Not Available
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-pyridone