accessibility menu, dialog, popup

UserName

NH2HNO

CAS RN 33630-99-8

IUPAC Name: 3-amino-1,2-dihydropyridin-2-one
Synonyms: 3-amino-1H-pyridin-2-one
Molecular Weight (g/mol): 110.12
Molecular Formula: C5H6N2O
InChi Key: VTSFNCCQCOEPKF-UHFFFAOYSA-N
SMILES: NC1=CC=CNC1=O
Percent Purity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (2)
  • (2)
  • (2)
Quantity 
  • (3)
  • (3)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (3)
  • (2)