missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-naphthol 98.0+%, TCI America™
$205.05 - $719.52
Chemical Identifiers
| CAS | 5417-63-0 |
|---|---|
| Molecular Formula | C10H9NO |
| Molecular Weight (g/mol) | 159.188 |
| MDL Number | MFCD00004113 |
| InChI Key | ZHVPTERSBUMMHK-UHFFFAOYSA-N |
| Synonym | 3-amino-2-naphthol, 2-naphthalenol, 3-amino, 2-amino-3-naphthol, 3-amino-2-hydroxynaphthalene, 2-naphthol, 3-amino, 2-amino-3-hydroxynaphthalene, unii-29bke27d17, 3-amino-naphthalen-2-ol, acmc-209ldx, 3-amino-2-naphthalenol |
| PubChem CID | 79449 |
| IUPAC Name | 3-aminonaphthalen-2-ol |
| SMILES | C1=CC=C2C=C(C(=CC2=C1)N)O |
Chemical Identifiers
| 5417-63-0 | |
| 159.188 | |
| ZHVPTERSBUMMHK-UHFFFAOYSA-N | |
| 79449 | |
| C1=CC=C2C=C(C(=CC2=C1)N)O |
| C10H9NO | |
| MFCD00004113 | |
| 3-amino-2-naphthol, 2-naphthalenol, 3-amino, 2-amino-3-naphthol, 3-amino-2-hydroxynaphthalene, 2-naphthol, 3-amino, 2-amino-3-hydroxynaphthalene, unii-29bke27d17, 3-amino-naphthalen-2-ol, acmc-209ldx, 3-amino-2-naphthalenol | |
| 3-aminonaphthalen-2-ol |
Specifications
| 5417-63-0 | |
| C10H9NO | |
| 1 g | |
| ZHVPTERSBUMMHK-UHFFFAOYSA-N | |
| 3-aminonaphthalen-2-ol | |
| 79449 | |
| ≥98.0% (GC,T) | |
| 3-Amino-2-naphthol |
| White-Yellow | |
| MFCD00004113 | |
| 3-amino-2-naphthol, 2-naphthalenol, 3-amino, 2-amino-3-naphthol, 3-amino-2-hydroxynaphthalene, 2-naphthol, 3-amino, 2-amino-3-hydroxynaphthalene, unii-29bke27d17, 3-amino-naphthalen-2-ol, acmc-209ldx, 3-amino-2-naphthalenol | |
| C1=CC=C2C=C(C(=CC2=C1)N)O | |
| 159.188 | |
| 159.19 | |
| Crystalline Powder |
Safety and Handling
TSCA : No