missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-naphthol 98.0+%, TCI America™

Catalog No. A15931G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A15931G 1 g
A15935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A15931G Supplier TCI America Supplier No. A15931G
Only null left

Chemical Identifiers

CAS 5417-63-0
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00004113
InChI Key ZHVPTERSBUMMHK-UHFFFAOYSA-N
Synonym 3-amino-2-naphthol, 2-naphthalenol, 3-amino, 2-amino-3-naphthol, 3-amino-2-hydroxynaphthalene, 2-naphthol, 3-amino, 2-amino-3-hydroxynaphthalene, unii-29bke27d17, 3-amino-naphthalen-2-ol, acmc-209ldx, 3-amino-2-naphthalenol
PubChem CID 79449
IUPAC Name 3-aminonaphthalen-2-ol
SMILES C1=CC=C2C=C(C(=CC2=C1)N)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 159.19
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-naphthol