missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-naphthoic Acid 97.0+%, TCI America™

Catalog No. A22581G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A22581G 1 g
A22585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A22581G Supplier TCI America Supplier No. A22581G

Chemical Identifiers

CAS 5959-52-4
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.198
MDL Number MFCD00004115
InChI Key XFXOLBNQYFRSLQ-UHFFFAOYSA-N
Synonym 3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
PubChem CID 22244
IUPAC Name 3-aminonaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N

Specifications

Melting Point 212°C
Color Yellow
Quantity 1 g
Formula Weight 187.20
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-naphthoic Acid
RTECSNumber QL1400000
TSCA Yes