missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-naphthoic acid, 97%

Catalog No. p-7047488
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1931203 1 g
AAL1931206 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1931203 Supplier Thermo Scientific Chemicals Supplier No. L1931203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 5959-52-4 | C11H9NO2 | 187.198 g/mol

3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-2-naphthoic acid is a dyes analytical reagent which is used as an intermediate for organic synthesis.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 5959-52-4
Molecular Formula C11H9NO2
MDL Number MFCD00004115
InChI Key XFXOLBNQYFRSLQ-UHFFFAOYSA-N
Synonym 3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
PubChem CID 22244
IUPAC Name 3-aminonaphthalene-2-carboxylic acid

Specifications

Melting Point ∽210°C (decomposition)
Quantity 1 g
Beilstein 744099
Merck Index 14,452
Solubility Information Soluble in alcohol and ether. Insoluble in water.
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N
Molecular Weight (g/mol) 187.198
Formula Weight 187.2
Percent Purity 97%
Chemical Name or Material 3-Amino-2-naphthoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 227-726-2
RTECSNumber QL1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only