missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-naphthoic acid, 85%, tech.

Catalog No. AC104140050
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC104140050 5 g Glass bottle
AC104140250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC104140050 Supplier Thermo Scientific Chemicals Supplier No. 104140050

CAS: 5959-52-4 | C11H9NO2 | 187.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 5959-52-4
Molecular Formula C11H9NO2
Molecular Weight (g/mol) 187.2
MDL Number MFCD00004115
InChI Key XFXOLBNQYFRSLQ-UHFFFAOYSA-N
Synonym 3-amino-2-naphthoic acid, 2-amino-3-naphthoic acid, 3-aminoisonaphthoic acid, 2-naphthalenecarboxylic acid, 3-amino, 2-amino-3-carboxynaphthalene, 2-naphthoic acid, 3-amino, 3-amino-2-naphthalenecarboxylic acid, unii-iv5z0phl5d, iv5z0phl5d, 3-amino-naphthalene-2-carboxylic acid
PubChem CID 22244
IUPAC Name 3-aminonaphthalene-2-carboxylic acid
SMILES C1=CC=C2C=C(C(=CC2=C1)C(=O)O)N

Specifications

CAS Min % 80.0
CAS Max % 100.0
Melting Point 210.0°C
Color Green to Green-Yellow
Infrared Spectrum Authentic
Assay Percent Range 85%
Packaging Glass bottle
Linear Formula H2NC10H6CO2H
Quantity 5 g
Beilstein 14, 535
Merck Index 15, 448
Formula Weight 187.2
Percent Purity 85%
Grade Technical
Physical Form Powder
Chemical Name or Material 3-Amino-2-naphthoic acid, 85%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove
EINECSNumber 227-726-2
RTECSNumber QL1400000
TSCA TSCA

RUO – Research Use Only