missing translation for 'onlineSavingsMsg'
Learn More

3'-Amino-2'-hydroxybiphenyl-3-carboxylic Acid 98.0+%, TCI America™

Catalog No. A2944200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A2944200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A2944200MG Supplier TCI America Supplier No. A2944200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 376592-93-7
Molecular Formula C13H11NO3
Molecular Weight (g/mol) 229.235
MDL Number MFCD10001467
InChI Key ZXLYSSHNDUXXIN-UHFFFAOYSA-N
Synonym 3-(3-Amino-2-hydroxyphenyl)benzoic Acid
PubChem CID 11492332
IUPAC Name 3-(3-amino-2-hydroxyphenyl)benzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C2=C(C(=CC=C2)N)O

Specifications

Melting Point 222°C
Quantity 200 mg
Formula Weight 229.24
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3′-Amino-2′-hydroxybiphenyl-3-carboxylic Acid