missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-hydroxy-5-nitrobenzenesulfonic Acid Monohydrate 98.0+%, TCI America™

Catalog No. p-7117786
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A21975G 5 g
A219725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A21975G Supplier TCI America Supplier No. A21975G
Only null left

Chemical Identifiers

CAS 96-67-3
Molecular Formula C6H6N2O6S
Molecular Weight (g/mol) 234.182
MDL Number MFCD00035758
InChI Key DQIVFTJHYKDOMZ-UHFFFAOYSA-N
PubChem CID 4312530
SMILES C1=C(C=C(C(=C1S(=O)(=O)O)O)N)[N+](=O)[O-]

Specifications

Quantity 5 g
Formula Weight 234.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-hydroxy-5-nitrobenzenesulfonic Acid Monohydrate
EINECSNumber (3)-2096
RTECSNumber OY2357500
TSCA Yes