Learn More
3-Amino-2-chloropyridine, 98+%
CAS: 6298-19-7 | C5H5ClN2 | 128.56 g/mol
Supplier: Thermo Scientific Chemicals A1583018
Description
It is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an intermediate of medicine and agricultural chemicals. It is involved in the Suzuki-Miyaura cross-coupling reactions of heteroaryl compounds.
Solubility
Soluble in methanol and water. (30 g/L).
Notes
Store at room temperature. Keep away from oxidizing agents.
Specifications
| 3-Amino-2-chloropyridine | |
| 76°C to 80°C | |
| 185°C (365°F) | |
| C5H5ClN2 | |
| 50 g | |
| 3-amino-2-chloropyridine, 2-chloro-3-aminopyridine, 2-chloro-3-pyridinamine, 3-pyridinamine, 2-chloro, 2-chloro-3-amino pyridine, 2-chloro-3-pyridylamine, 2-chloro-3-amino-pyridine, 2-chloro-pyridin-3-ylamine, 2-chloro-3-pyridinylamine, 3-amino-2-chloro-pyridine | |
| MEQBJJUWDCYIAB-UHFFFAOYSA-N | |
| 2-chloropyridin-3-amine | |
| 80528 | |
| ≥98% |
| 6298-19-7 | |
| 130°C to 134°C (22 mmHg) | |
| Unpleasant | |
| MFCD00006238 | |
| 109814 | |
| Soluble in methanol and water. (30g/L). | |
| NC1=CC=CN=C1Cl | |
| 128.56 | |
| 128.56 |
Chemical Identifiers
| 6298-19-7 | |
| 128.56 | |
| MEQBJJUWDCYIAB-UHFFFAOYSA-N | |
| 80528 | |
| NC1=CC=CN=C1Cl |
| C5H5ClN2 | |
| MFCD00006238 | |
| 3-amino-2-chloropyridine, 2-chloro-3-aminopyridine, 2-chloro-3-pyridinamine, 3-pyridinamine, 2-chloro, 2-chloro-3-amino pyridine, 2-chloro-3-pyridylamine, 2-chloro-3-amino-pyridine, 2-chloro-pyridin-3-ylamine, 2-chloro-3-pyridinylamine, 3-amino-2-chloro-pyridine | |
| 2-chloropyridin-3-amine |
Safety and Handling
GHS H Statement
H312-H315-H319-H335
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 228-572-9
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only