missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloro-4-(trifluoromethyl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH3302603
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3302603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3302603 Supplier Thermo Scientific Chemicals Supplier No. H3302603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 166770-70-3 | Molecular Formula: C6H4ClF3N2 | Molecular Weight: 196.557 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 166770-70-3
Molecular Formula C6H4ClF3N2
Molecular Weight (g/mol) 196.557
MDL Number MFCD09863900
InChI Key KFMUXCNJJIWKRE-UHFFFAOYSA-N
Synonym 2-chloro-3-amino-4-trifluoromethyl pyridine, 2-chloro-4-trifluoromethyl pyridin-3-amine, 2-chloro-4-trifluoromethyl-3-pyridinamine, 3-amino-2-chloro-4-trifluoromethyl pyridine, 2-chloro-4-trifluoromethyl-pyridin-3-ylamine
PubChem CID 19851909
IUPAC Name 2-chloro-4-(trifluoromethyl)pyridin-3-amine
SMILES C1=CN=C(C(=C1C(F)(F)F)N)Cl

Specifications

Melting Point 56°C to 59°C
Quantity 1 g
UN Number UN2811
Formula Weight 196.56
Percent Purity 97%
Chemical Name or Material 3-Amino-2-chloro-4-(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only