accessibility menu, dialog, popup

UserName

CAS RN 166770-70-3

IUPAC Name: 2-chloro-4-(trifluoromethyl)pyridin-3-amine
Synonyms: 2-chloro-4-(trifluoromethyl)pyridin-3-amine
Molecular Weight (g/mol): 196.56
Molecular Formula: C6H4ClF3N2
InChi Key: KFMUXCNJJIWKRE-UHFFFAOYSA-N
SMILES: NC1=C(C=CN=C1Cl)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results