missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2-chloro-4-methylpyridine 98.0+%, TCI America™

Catalog No. A18555G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A18555G 5 g
A185525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A18555G Supplier TCI America Supplier No. A18555G
Only null left

Chemical Identifiers

CAS 133627-45-9
Molecular Formula C6H7ClN2
Molecular Weight (g/mol) 142.59
MDL Number MFCD00673152
InChI Key UOBCYTOUXLAABU-UHFFFAOYSA-N
Synonym 3-amino-2-chloro-4-methylpyridine, 2-chloro-3-amino-4-methylpyridine, 3-amino-2-chloro-4-picoline, 2-chloro-3-amino-4-methyl pyridine, 2-chloro-4-methyl-pyridin-3-ylamine, 2-chloro-3-amino-4-picoline, 2-chloro-4-methyl-3-pyridinamine, 3-pyridinamine, 2-chloro-4-methyl, 2-chloro-4-methyl-3-pyridylamine, 3-amino-2-chloro-4-methyl pyridine
PubChem CID 2756387
IUPAC Name 2-chloro-4-methylpyridin-3-amine
SMILES CC1=CC=NC(Cl)=C1N

Specifications

Melting Point 68°C
Quantity 5 g
Formula Weight 142.59
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-2-chloro-4-methylpyridine