accessibility menu, dialog, popup

UserName

CH3NClH2N

CAS RN 133627-45-9

IUPAC Name: 2-chloro-4-methylpyridin-3-amine
Synonyms: 2-chloro-4-methylpyridin-3-amine
Molecular Weight (g/mol): 142.59
Molecular Formula: C6H7ClN2
InChi Key: UOBCYTOUXLAABU-UHFFFAOYSA-N
SMILES: CC1=CC=NC(Cl)=C1N
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results