missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2-bromo-5-chloropyridine 98.0+%, TCI America™
$89.48 - $264.70
Chemical Identifiers
| CAS | 90902-83-3 |
|---|---|
| Molecular Formula | C5H4BrClN2 |
| Molecular Weight (g/mol) | 207.455 |
| MDL Number | MFCD07368976 |
| InChI Key | AZOXQBMBDRLSCQ-UHFFFAOYSA-N |
| PubChem CID | 2769640 |
| IUPAC Name | 2-bromo-5-chloropyridin-3-amine |
| SMILES | C1=C(C(=NC=C1Cl)Br)N |
Chemical Identifiers
| 90902-83-3 | |
| 207.455 | |
| AZOXQBMBDRLSCQ-UHFFFAOYSA-N | |
| 2-bromo-5-chloropyridin-3-amine |
| C5H4BrClN2 | |
| MFCD07368976 | |
| 2769640 | |
| C1=C(C(=NC=C1Cl)Br)N |
Specifications
| 90902-83-3 | |
| C5H4BrClN2 | |
| 5 g | |
| C1=C(C(=NC=C1Cl)Br)N | |
| 207.455 | |
| 207.46 | |
| Crystalline Powder |
| 144°C | |
| MFCD07368976 | |
| AZOXQBMBDRLSCQ-UHFFFAOYSA-N | |
| 2-bromo-5-chloropyridin-3-amine | |
| 2769640 | |
| ≥98.0% (GC) | |
| 3-Amino-2-bromo-5-chloropyridine |
Safety and Handling
TSCA : No