accessibility menu, dialog, popup

UserName

CAS RN 90902-83-3

NClBrNH2

CAS RN 90902-83-3

IUPAC Name: 2-bromo-5-chloropyridin-3-amine
Synonyms: 2-bromo-5-chloropyridin-3-amine
Molecular Weight (g/mol): 207.46
Molecular Formula: C5H4BrClN2
InChi Key: AZOXQBMBDRLSCQ-UHFFFAOYSA-N
SMILES: NC1=CC(Cl)=CN=C1Br
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results