missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2,5-dichlorobenzoic Acid 98.0+%, TCI America™
Supplier: TCI America A15375G
Specifications
| 3-Amino-2,5-dichlorobenzoic Acid | |
| C7H5Cl2NO2 | |
| 5 g | |
| HSSBORCLYSCBJR-UHFFFAOYSA-N | |
| 3-amino-2,5-dichlorobenzoic acid | |
| 8630 | |
| 206.02 | |
| Crystalline Powder |
| 133-90-4 | |
| MFCD00065093 | |
| chloramben, ambiben, amiben, amibin, amoben, vegiben, ornamental weeder, amiben ds, ornamental weeder 4g, benzoic acid, 3-amino-2,5-dichloro | |
| C1=C(C=C(C(=C1N)Cl)C(=O)O)Cl | |
| 206.022 | |
| CHEBI:82183 | |
| ≥98.0% (T) |
Chemical Identifiers
| 133-90-4 | |
| 206.022 | |
| HSSBORCLYSCBJR-UHFFFAOYSA-N | |
| 8630 | |
| 3-amino-2,5-dichlorobenzoic acid |
| C7H5Cl2NO2 | |
| MFCD00065093 | |
| chloramben, ambiben, amiben, amibin, amoben, vegiben, ornamental weeder, amiben ds, ornamental weeder 4g, benzoic acid, 3-amino-2,5-dichloro | |
| CHEBI:82183 | |
| C1=C(C=C(C(=C1N)Cl)C(=O)O)Cl |
Safety and Handling
RTECSNumber : DG1925000
TSCA : Yes