missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Amino-2,4-difluorobenzeneboronic acid pinacol ester, 96%
CAS: 1315692-91-1 | C12H16BF2NO2 | 255.072 g/mol
$293.04 - $840.29
Chemical Identifiers
| CAS | 1315692-91-1 |
|---|---|
| Molecular Formula | C12H16BF2NO2 |
| Molecular Weight (g/mol) | 255.072 |
| MDL Number | MFCD22419270 |
| InChI Key | MQGLBRAIKSVSIT-UHFFFAOYSA-N |
| Synonym | 3-amino-2,4-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2,4-difluorobenzeneboronicacidpinacolester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline |
| PubChem CID | 99738339 |
| IUPAC Name | 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
| SMILES | B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)N)F |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAH59033MD
|
Thermo Scientific Chemicals
H59033MD |
250 mg |
Each for $293.04
|
|
|||||
|
AAH5903303
|
Thermo Scientific Chemicals
H5903303 |
1 g |
Each for $840.29
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 1315692-91-1 | |
| 255.072 | |
| MQGLBRAIKSVSIT-UHFFFAOYSA-N | |
| 99738339 | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)N)F |
| C12H16BF2NO2 | |
| MFCD22419270 | |
| 3-amino-2,4-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2,4-difluorobenzeneboronicacidpinacolester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline | |
| 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline |
Specifications
| 1315692-91-1 | |
| C12H16BF2NO2 | |
| 250 mg | |
| MQGLBRAIKSVSIT-UHFFFAOYSA-N | |
| 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline | |
| 99738339 | |
| 96% |
| 75°C to 77°C | |
| MFCD22419270 | |
| 3-amino-2,4-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2,4-difluorobenzeneboronicacidpinacolester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline | |
| B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)N)F | |
| 255.072 | |
| 255.07 | |
| 3-Amino-2,4-difluorobenzeneboronic acid pinacol ester |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only