missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,4-difluorobenzeneboronic acid pinacol ester, 96%

Catalog No. aah5903303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5903303 1 g
AAH59033MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5903303 Supplier Thermo Scientific Chemicals Supplier No. H5903303
Only null left

CAS: 1315692-91-1 | C12H16BF2NO2 | 255.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1315692-91-1
Molecular Formula C12H16BF2NO2
Molecular Weight (g/mol) 255.072
MDL Number MFCD22419270
InChI Key MQGLBRAIKSVSIT-UHFFFAOYSA-N
Synonym 3-amino-2,4-difluorobenzeneboronic acid pinacol ester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl aniline, 3-amino-2,4-difluorobenzeneboronicacidpinacolester, 2,6-difluoro-3-4,4,5,5-tetramethyl-1,3,2-dioxaborolane-2-yl aniline
PubChem CID 99738339
IUPAC Name 2,6-difluoro-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline
SMILES B1(OC(C(O1)(C)C)(C)C)C2=C(C(=C(C=C2)F)N)F

Specifications

Melting Point 75°C to 77°C
Quantity 1 g
Formula Weight 255.07
Percent Purity 96%
Chemical Name or Material 3-Amino-2,4-difluorobenzeneboronic acid pinacol ester
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only