missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,4,6-triiodobenzoic acid, 99% (dry wt.), water <4%

Catalog No. AAB2340514
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2340514 25 g
AAB2340506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2340514 Supplier Thermo Scientific Chemicals Supplier No. B2340514
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3119-15-1 | C7H3I3NNaO2 | 536.81 g/mol

3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Amino-2,4,6-triiodobenzoic acid is used to prepare polymeric hydrogel based particles used in vascular embolization. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Insoluble in water.

Notes
Protect from exposure to light. Store in the dark. Incompatible with light and oxidizing agents. Store at room temperature.

Chemical Identifiers

CAS 3119-15-1
Molecular Formula C7H3I3NNaO2
Molecular Weight (g/mol) 536.81
MDL Number MFCD00007681
InChI Key CTXRMWPLTDAHOR-UHFFFAOYSA-M
Synonym unii-99h77d8mtd, 3-amino-2,4,6-triiodo-benzoic acid, acido 3-amino-2,4,6-triiodobenzoico italian, benzoic acid, 3-amino-2,4,6-triiodo, 3-amino-2,4,6-triiodo-benzoicacid, acido 3-amino-2,4,6-triiodobenzoico, pubchem4987, acmc-1co9w, 4-14-00-01118 beilstein handbook reference, 2,4,6-triiodo-3-aminobenzoic acid
PubChem CID 18387
IUPAC Name 3-amino-2,4,6-triiodobenzoic acid
SMILES [Na+].NC1=C(I)C=C(I)C(C([O-])=O)=C1I

Specifications

Melting Point ∼196°C (decomposition)
Quantity 25 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 514.83
Percent Purity 99%
Assay (dry wt.), may cont. up to 1 mole of water
Chemical Name or Material 3-Amino-2,4,6-triiodobenzoic acid
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 221-493-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only