missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,2-dimethyl-1-propanol 97.0+%, TCI America™

Catalog No. p-7117127
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
A09145G 5 g
A091425G 25 g
2 options

Catalog No. A09145G

Supplier: TCI America A09145G

Only null left

Chemical Identifiers

CAS 26734-09-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00059847
InChI Key FNVOFDGAASRDQY-UHFFFAOYSA-N
Synonym 3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
PubChem CID 117326
IUPAC Name 3-amino-2,2-dimethylpropan-1-ol
SMILES CC(C)(CN)CO

Specifications

Color White
Quantity 5 g
UN Number 3263
Formula Weight 103.17
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol