accessibility menu, dialog, popup

UserName

CH3H3CNH2HO

CAS RN 26734-09-8

IUPAC Name: 3-amino-2,2-dimethylpropan-1-ol
Synonyms: 3-amino-2,2-dimethylpropan-1-ol
Molecular Weight (g/mol): 103.17
Molecular Formula: C5H13NO
InChi Key: FNVOFDGAASRDQY-UHFFFAOYSA-N
SMILES: CC(C)(CN)CO
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)