missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-2,2-dimethyl-1-propanol, 95%

Catalog No. AAL1042903
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1042903 1 g
AAL1042906 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1042903 Supplier Thermo Scientific Chemicals Supplier No. L1042903
Only null left

CAS: 26734-09-8 | C5H13NO | 103.165 g/mol

Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Reatant in the synthesis of cyclic carbamates, cryptophycin analogues. Neopentanolamine with ammonia and hydrogen yeilds neopentyldiamine.

Solubility
Soluble in Dichloromethane, Chloroforn, Ether and Methanol

Notes
Hygroscopic,Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture.

Chemical Identifiers

CAS 26734-09-8
Molecular Formula C5H13NO
Molecular Weight (g/mol) 103.165
MDL Number MFCD00059847
InChI Key FNVOFDGAASRDQY-UHFFFAOYSA-N
Synonym 3-amino-2,2-dimethyl-1-propanol, neopentanolamine, 1-propanol, 3-amino-2,2-dimethyl, 3-amino-2,2-dimethylpropanol, 2,2-dimethyl-3-aminopropanol, 1-amino-2,2-dimethyl-3-propanol, 3-amino-2,2-dimethyl-propan-1-ol, neo-pentanolamine, acmc-1cdms, ksc496k6d
PubChem CID 117326
IUPAC Name 3-amino-2,2-dimethylpropan-1-ol
SMILES CC(C)(CN)CO

Specifications

Melting Point ∼70°C
Quantity 1 g
Beilstein 1731900
Sensitivity Hygroscopic
Solubility Information Soluble in Dichloromethane,Chloroforn,Ether and Methanol
Formula Weight 103.17
Percent Purity 95%
Chemical Name or Material 3-Amino-2,2-dimethyl-1-propanol
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 247-948-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only