Learn More
3-Amino-1H-1,2,4-triazole, 95%
CAS: 61-82-5 | C2H4N4 | 84.08 g/mol
$187.88 - $187.88
Chemical Identifiers
| CAS | 61-82-5 |
|---|---|
| Molecular Formula | C2H4N4 |
| Molecular Weight (g/mol) | 84.08 |
| MDL Number | MFCD00005230,MFCD00053362 |
| InChI Key | KLSJWNVTNUYHDU-UHFFFAOYSA-N |
| Synonym | 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox |
| PubChem CID | 1639 |
| ChEBI | CHEBI:40036 |
| IUPAC Name | 1H-1,2,4-triazol-5-amine |
| SMILES | NC1=NC=NN1 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC264571000
|
Thermo Scientific Chemicals
264571000 |
100 g | Glass bottle |
Each for $187.88
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 61-82-5 | |
| 84.08 | |
| KLSJWNVTNUYHDU-UHFFFAOYSA-N | |
| 1639 | |
| 1H-1,2,4-triazol-5-amine |
| C2H4N4 | |
| MFCD00005230,MFCD00053362 | |
| 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox | |
| CHEBI:40036 | |
| NC1=NC=NN1 |
Specifications
| 61-82-5 | |
| 100.0 | |
| White or Yellow | |
| 95% | |
| C2H4N4 | |
| 100 g | |
| 15, 485 | |
| Solubility in water: 280g/L (20°C). Other solubilities: 520g/L (50°C),sparingly soluble in chloroform,dichloromethane,and acetonitrile,slightly soluble in ethylacetate,pract.insol. in diethylether,hydrocarbons,acetone | |
| NC1=NC=NN1 | |
| 84.08 | |
| CHEBI:40036 | |
| 95% | |
| 3-Amino-1H-1, 2, 4-triazole, 95% |
| 94.0 | |
| 149°C to 156°C | |
| Authentic | |
| Glass bottle | |
| MFCD00005230,MFCD00053362 | |
| 26, 137 | |
| 3-amino-1,2,4-triazole, amitrole, aminotriazole, 4h-1,2,4-triazol-3-amine, 1h-1,2,4-triazol-3-amine, amitrol, amerol, amizol, emisol, vorox | |
| KLSJWNVTNUYHDU-UHFFFAOYSA-N | |
| 1H-1,2,4-triazol-5-amine | |
| 1639 | |
| 84.08 | |
| Powder or Flakes |
Safety and Handling
GHS H Statement:
May cause damage to organs through prolonged or repeated exposure.
Toxic to aquatic life with long lasting effects.
Suspected of damaging the unborn child.
GHS P Statement:
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 200-521-5
RTECSNumber : XZ3850000
TSCA : TSCA
RUO – Research Use Only